missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)-3,5-dimethylisoxazole 95.0+%, TCI America™
$108.11 - $328.08
Chemical Identifiers
| CAS | 19788-37-5 |
|---|---|
| Molecular Formula | C6H8ClNO |
| Molecular Weight (g/mol) | 145.586 |
| MDL Number | MFCD00003154 |
| InChI Key | NIFAUKBQIAURIM-UHFFFAOYSA-N |
| Synonym | 4-chloromethyl-3,5-dimethylisoxazole, isoxazole, 4-chloromethyl-3,5-dimethyl, 4-chloromethyl-3,5-dimethyl-1,2-oxazole, 4-chloromethyl-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-chloromethylisoxazole, 4-chloromethyl-3,5-dimethyl isoxazole, pubchem8667, acmc-1c8qy, akos bc-3064 |
| PubChem CID | 88246 |
| IUPAC Name | 4-(chloromethyl)-3,5-dimethyl-1,2-oxazole |
| SMILES | CC1=C(C(=NO1)C)CCl |
Chemical Identifiers
| 19788-37-5 | |
| 145.586 | |
| NIFAUKBQIAURIM-UHFFFAOYSA-N | |
| 88246 | |
| CC1=C(C(=NO1)C)CCl |
| C6H8ClNO | |
| MFCD00003154 | |
| 4-chloromethyl-3,5-dimethylisoxazole, isoxazole, 4-chloromethyl-3,5-dimethyl, 4-chloromethyl-3,5-dimethyl-1,2-oxazole, 4-chloromethyl-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-chloromethylisoxazole, 4-chloromethyl-3,5-dimethyl isoxazole, pubchem8667, acmc-1c8qy, akos bc-3064 | |
| 4-(chloromethyl)-3,5-dimethyl-1,2-oxazole |
Specifications
| 19788-37-5 | |
| C6H8ClNO | |
| 1 g | |
| NIFAUKBQIAURIM-UHFFFAOYSA-N | |
| 4-(chloromethyl)-3,5-dimethyl-1,2-oxazole | |
| 88246 | |
| ≥95.0% (GC) | |
| 4-(Chloromethyl)-3,5-dimethylisoxazole |
| 88°C | |
| MFCD00003154 | |
| 4-chloromethyl-3,5-dimethylisoxazole, isoxazole, 4-chloromethyl-3,5-dimethyl, 4-chloromethyl-3,5-dimethyl-1,2-oxazole, 4-chloromethyl-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-chloromethylisoxazole, 4-chloromethyl-3,5-dimethyl isoxazole, pubchem8667, acmc-1c8qy, akos bc-3064 | |
| CC1=C(C(=NO1)C)CCl | |
| 145.586 | |
| 145.59 | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator