missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)-3,5-dimethylisoxazole 95.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 88°C |
| Quantity | 1 g |
| Formula Weight | 145.59 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-(Chloromethyl)-3,5-dimethylisoxazole |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Refrigerator |