missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromethyl-2-methylthiazole hydrochloride, 98%

Catalog No. p-7022228
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1490206 5 g
AAA1490214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1490206 Supplier Thermo Scientific Chemicals Supplier No. A1490206
Only null left

CAS: 77470-53-2 | C5H7Cl2NS | 184.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77470-53-2
Molecular Formula C5H7Cl2NS
Molecular Weight (g/mol) 184.08
MDL Number MFCD00067725
InChI Key YGKDISJLDVGNOR-UHFFFAOYSA-N
Synonym 4-chloromethyl-2-methylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-methylthiazole hcl, 4-chloromethyl-2-methyl thiazole hydrochloride, 2-methyl-4-chloromethyl thiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole, chloride, 2-methyl-4-chloromethylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hcl
PubChem CID 2734203
SMILES Cl.CC1=NC(CCl)=CS1

Specifications

Melting Point ∼165°C (decomposition)
Quantity 5 g
UN Number UN3261
Beilstein 108603
IUPAC Name 4-(chloromethyl)-2-methyl-1,3-thiazole hydrochloride
Formula Weight 184.09
Percent Purity 98%
Chemical Name or Material 4-Chloromethyl-2-methylthiazole hydrochloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only