missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-2-methyl-1,3-thiazole hydrochloride , Tech., Thermo Scientific™

Catalog No. p-4509946
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD09497CB 250mg
CD09497DA 1g
CD09497DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CD09497CB Supplier Thermo Scientific Chemicals Supplier No. CD09497CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77470-53-2
Molecular Formula C5H7Cl2NS
Molecular Weight (g/mol) 184.08
MDL Number MFCD00067725
InChI Key YGKDISJLDVGNOR-UHFFFAOYSA-N
Synonym 4-chloromethyl-2-methylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-methylthiazole hcl, 4-chloromethyl-2-methyl thiazole hydrochloride, 2-methyl-4-chloromethyl thiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole, chloride, 2-methyl-4-chloromethylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hcl
PubChem CID 2734203
IUPAC Name 4-(chloromethyl)-2-methyl-1,3-thiazole;hydrochloride
SMILES Cl.CC1=NC(CCl)=CS1

Specifications

Packaging Amber glass bottle
Quantity 250mg
Formula Weight 184.09
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(Chloromethyl)-2-methyl-1, 3-thiazole hydrochloride