missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-1-(2-furylmethyl)piperidine hydrochloride, 95%, Thermo Scientific™

Catalog No. CC68574CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC68574CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC68574CB Supplier Thermo Scientific Chemicals Supplier No. CC68574CB

Chemical Identifiers

CAS 944450-92-4
Molecular Formula C11H17Cl2NO
Molecular Weight (g/mol) 250.163
MDL Number MFCD10700059
InChI Key SRRFZUHMIKVVPV-UHFFFAOYSA-N
Synonym 4-chloromethyl-1-2-furylmethyl piperidine hydrochloride, 4-chloromethyl-1-furan-2-ylmethyl piperidine hydrochloride, 2-4-chloromethyl piperidyl methyl furan, chloride, 4-chloromethyl-1-furan-2-yl methyl piperidine-hydrogen chloride 1/1
PubChem CID 43811048
IUPAC Name 4-(chloromethyl)-1-(furan-2-ylmethyl)piperidine;hydrochloride
SMILES C1CN(CCC1CCl)CC2=CC=CO2.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 250.17
Percent Purity 95%
Chemical Name or Material 4-(Chloromethyl)-1-(2-furylmethyl)piperidine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.