missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromandelic acid, 98%

Catalog No. p-7027481
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2426406 5 g
AAB2426414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2426406 Supplier Thermo Scientific Chemicals Supplier No. B2426406
Only null left

CAS: 492-86-4 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 492-86-4
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00042724
InChI Key BWSFWXSSALIZAU-UHFFFAOYSA-N
Synonym 4-chloromandelic acid, 2-4-chlorophenyl-2-hydroxyacetic acid, p-chloromandelic acid, p-chloro mandelic acid, 4-chloro-dl-mandelic acid, 4-chloromandelic acid, pract., benzeneacetic acid, 4-chloro-.alpha.-hydroxy, dl-4-chloromandelic acid, mandelic acid, p-chloro, 4-chlorophenyl hydroxy acetic acid
PubChem CID 10299
IUPAC Name 2-(4-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)O)Cl

Specifications

Melting Point 117°C to 122°C
Quantity 5 g
Beilstein 1567759
Formula Weight 186.6
Percent Purity 98%
Chemical Name or Material 4-Chloromandelic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-764-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only