missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorocinnamic Acid 98.0+%, TCI America™

Catalog No. p-7124248
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C014925G 25 g
C0149500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C014925G Supplier TCI America Supplier No. C014925G
Only null left

Chemical Identifiers

CAS 1615-02-7
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.60
MDL Number MFCD00004396
InChI Key GXLIFJYFGMHYDY-ZZXKWVIFSA-N
Synonym 4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid
PubChem CID 637797
ChEBI CHEBI:61116
IUPAC Name (2E)-3-(4-chlorophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=C(Cl)C=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 182.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorocinnamic Acid
RTECSNumber GD8490000
TSCA No