missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyryl Chloride 98.0+%, TCI America™

Catalog No. C082425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C082425ML 25 mL
C0824500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C082425ML Supplier TCI America Supplier No. C082425ML

Chemical Identifiers

CAS 4635-59-0
Molecular Formula C4H6Cl2O
Molecular Weight (g/mol) 140.991
MDL Number MFCD00000754
InChI Key CDIIZULDSLKBKV-UHFFFAOYSA-N
Synonym 4-chlorobutyryl chloride, butanoyl chloride, 4-chloro, 4-chlorobutanoic acid chloride, 4-chlorobutyric acid chloride, 4-chlorobutyroyl chloride, gamma-chlorobutyryl chloride, 4-chlorobutyrylchloride, gamma-chlorobutyroyl chloride, unii-3663u0vkri, butyryl chloride, 4-chloro
PubChem CID 78370
IUPAC Name 4-chlorobutanoyl chloride
SMILES C(CC(=O)Cl)CCl

Specifications

Color Yellow
Boiling Point 174°C
Quantity 25 mL
UN Number 2922
Formula Weight 140.99
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyryl Chloride
EINECSNumber (2)-3787
RTECSNumber EM1406000
TSCA Yes