missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyronitrile 97.0+%, TCI America™

Catalog No. p-7124687
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
C092525ML 25 mL
C0925500ML 500 mL
2 options

Catalog No. C092525ML

Supplier: TCI America C092525ML

Only null left

Chemical Identifiers

CAS 628-20-6
Molecular Formula C4H6ClN
Molecular Weight (g/mol) 103.55
MDL Number MFCD00001972
InChI Key ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
Synonym 4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
PubChem CID 12336
IUPAC Name 4-chlorobutanenitrile
SMILES ClCCCC#N

Specifications

Color Yellow
Boiling Point 194°C
Quantity 25 mL
UN Number 3276
Formula Weight 103.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyronitrile
EINECSNumber (9)-1905
RTECSNumber ET9020000
TSCA Yes