missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyronitrile 97.0+%, TCI America™

Catalog No. C092525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C092525ML 25 mL
C0925500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C092525ML Supplier TCI America Supplier No. C092525ML

Chemical Identifiers

CAS 628-20-6
Molecular Formula C4H6ClN
Molecular Weight (g/mol) 103.55
MDL Number MFCD00001972
InChI Key ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
Synonym 4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
PubChem CID 12336
IUPAC Name 4-chlorobutanenitrile
SMILES ClCCCC#N

Specifications

Color Yellow
Boiling Point 194°C
Quantity 25 mL
UN Number 3276
Formula Weight 103.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyronitrile
EINECSNumber (9)-1905
RTECSNumber ET9020000
TSCA Yes