missing translation for 'onlineSavingsMsg'
Learn More

4-chlorobutyronitrile, 97%

Catalog No. AC329945000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329945000 500 g Glass bottle
AC329941000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC329945000 Supplier Thermo Scientific Chemicals Supplier No. 329945000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 628-20-6 | C4H6ClN | 103.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 628-20-6
Molecular Formula C4H6ClN
Molecular Weight (g/mol) 103.55
MDL Number MFCD00001972
InChI Key ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
Synonym 4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
PubChem CID 12336
IUPAC Name 4-chlorobutanenitrile
SMILES ClCCCC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 195°C to 197°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4420 to 1.444
Quantity 500 g
Beilstein 02, 278
Specific Gravity 1.08
Solubility Information Solubility in water: immiscible
Formula Weight 103.55
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-chlorobutyronitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-031-6

RUO – Research Use Only