missing translation for 'onlineSavingsMsg'
Learn More

4-chlorobutyronitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 329940050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC329940050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

628-20-6
103.55
ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
12336
ClCCCC#N
C4H6ClN
MFCD00001972
4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
4-chlorobutanenitrile

Specifications

4-chlorobutyronitrile
96.0
1.0800g/mL
85°C
96% min. (GC)
C4H6ClN
5g
02, 278
4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
4-chlorobutanenitrile
12336
97%
628-20-6
100.0
195°C to 197°C
Authentic
Glass bottle
1.4420 to 1.444
MFCD00001972
1.08
Solubility in water: immiscible
ClCCCC#N
103.55
103.55
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-031-6

SDS
Documents

Documents

Promotions

Promotions