missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyronitrile, 97%

Catalog No. aaa189420b
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1000 g
250 g
50 g
Catalog No. Quantity
AAA189420B 1000 g
AAA1894218 50 g
AAA1894230 250 g
3 options

Catalog No. AAA189420B

Supplier: Thermo Scientific Chemicals A189420B

Only null left

CAS: 628-20-6 | C4H6ClN | 103.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 628-20-6
Molecular Formula C4H6ClN
Molecular Weight (g/mol) 103.55
MDL Number MFCD00001972
InChI Key ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
Synonym 4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
PubChem CID 12336
IUPAC Name 4-chlorobutanenitrile
SMILES ClCCCC#N

Specifications

Density 1.084
Boiling Point 195°C to 197°C
Flash Point 85°C (185°F)
Refractive Index 1.444
Linear Formula Cl(CH2)3CN
Quantity 1000 g
UN Number UN3276
Beilstein 1633582
Formula Weight 103.55
Percent Purity 97%
Chemical Name or Material 4-Chlorobutyronitrile
Show More Show Less
Hazard Category H227-H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 211-031-6
RTECSNumber ET9020000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only