missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzylzinc chloride, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58532AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58532AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58532AD Supplier Thermo Scientific Chemicals Supplier No. H58532AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 149923-10-4 | C7H6Cl2Zn | 226.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzylzinc chloride used as a intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a

Chemical Identifiers

CAS 149923-10-4
Molecular Formula C7H6Cl2Zn
Molecular Weight (g/mol) 226.41
MDL Number MFCD01319882
InChI Key GIUOJMHOGPURPS-UHFFFAOYSA-M
Synonym 4-chlorobenzylzinc chloride solution, 4-chlorobenzylzinc chloride, chloro 4-chlorophenyl methyl zinc, 4-chlorobenzylzinc chloride solution, 0.5 m in thf, 4-chlorobenzylzinc chloride 0.5 m in tetrahydrofuran, 4-chlorobenzylzinc chloride, 0.5m in thf, packaged under arg, 4-chlorobenzylzinc chloride,0.5m soluti onin tetrahydrofuran, 4-chlorobenzylzinc chloride, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 3451123
IUPAC Name 1-chloro-4-methanidylbenzene;chlorozinc(1+)
SMILES Cl[Zn+].[CH2-]C1=CC=C(Cl)C=C1

Specifications

Density 0.96
Quantity 50 mL
UN Number UN3399
Solubility Information Reacts with water.
Formula Weight 226.41
Chemical Name or Material 4-Chlorobenzylzinc chloride, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H302-H314-H335-H351
Hazard Statement GHS H Statement
H225-H261-H314
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only