missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzylamine 98.0+%, TCI America™

Catalog No. c109025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C109025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C109025G Supplier TCI America Supplier No. C109025G
Only null left

Chemical Identifiers

CAS 104-86-9
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00008121
InChI Key YMVFJGSXZNNUDW-UHFFFAOYSA-N
Synonym 4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218
PubChem CID 66036
IUPAC Name (4-chlorophenyl)methanamine
SMILES C1=CC(=CC=C1CN)Cl

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 g
UN Number 2735
Formula Weight 141.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzylamine