missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl isothiocyanate, 97%

Catalog No. AAL0957906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0957906 5 g
AAL0957903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0957906 Supplier Thermo Scientific Chemicals Supplier No. L0957906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3694-45-9 | C8H6ClNS | 183.653 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3694-45-9
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.653
MDL Number MFCD00041116
InChI Key DEHXIHUIYSXZNH-UHFFFAOYSA-N
Synonym 4-chlorobenzyl isothiocyanate, 1-chloro-4-isothiocyanatomethyl benzene, p-chlorobenzyl isothiocyanate, benzene, 1-chloro-4-isothiocyanatomethyl, isothiocyanic acid, p-chlorobenzyl ester, 4-chlorobenzylisothiocyanate, benzene, 1-chloro-4-isothiocyanatomethyl-9ci, acmc-1cng5, wln: scn1r dg, p-chlorobenzylisothiocyanate
PubChem CID 19408
IUPAC Name 1-chloro-4-(isothiocyanatomethyl)benzene
SMILES C1=CC(=CC=C1CN=C=S)Cl

Specifications

Density 1.27
Quantity 5 g
Odor Irritating
Refractive Index 1.6145
UN Number UN2922
Beilstein 2085917
Boiling Point 169°C to 170°C (25 mmHg)
Formula Weight 183.66
Percent Purity 97%
Chemical Name or Material 4-Chlorobenzyl isothiocyanate
Hazard Category H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
RTECSNumber NX8470000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only