missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoylacetonitrile, 98%

CAS: 4640-66-8 | C9H6ClNO | 179.603 g/mol

Supplier:  Thermo Scientific Chemicals A1236414

 View more versions of this product

Catalog No. AAA1236414


Only null left
Explore available promotions

Description

Description

Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical Intermediates.

Solubility
Soluble in Dichloromethane, Ether, Ethyl Acetate and Methanol

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

4-Chlorobenzoylacetonitrile
128°C to 132°C
MFCD00051625
743368
Soluble in Dichloromethane,Ether,Ethyl Acetate and Methanol
C1=CC(=CC=C1C(=O)CC#N)Cl
179.603
179.61
4640-66-8
C9H6ClNO
25 g
4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag
JYOUFPNYTOFCSJ-UHFFFAOYSA-N
3-(4-chlorophenyl)-3-oxopropanenitrile
138308
98%

Chemical Identifiers

4640-66-8
179.603
JYOUFPNYTOFCSJ-UHFFFAOYSA-N
138308
C1=CC(=CC=C1C(=O)CC#N)Cl
C9H6ClNO
MFCD00051625
4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag
3-(4-chlorophenyl)-3-oxopropanenitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only