missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Chlorobenzoyl)acetonitrile 98.0+%, TCI America™
$748.89 - $2987.87
Chemical Identifiers
| CAS | 4640-66-8 |
|---|---|
| Molecular Formula | C9H6ClNO |
| Molecular Weight (g/mol) | 179.603 |
| MDL Number | MFCD00051625 |
| InChI Key | JYOUFPNYTOFCSJ-UHFFFAOYSA-N |
| Synonym | 4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag |
| PubChem CID | 138308 |
| IUPAC Name | 3-(4-chlorophenyl)-3-oxopropanenitrile |
| SMILES | C1=CC(=CC=C1C(=O)CC#N)Cl |
Chemical Identifiers
| 4640-66-8 | |
| 179.603 | |
| JYOUFPNYTOFCSJ-UHFFFAOYSA-N | |
| 138308 | |
| C1=CC(=CC=C1C(=O)CC#N)Cl |
| C9H6ClNO | |
| MFCD00051625 | |
| 4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag | |
| 3-(4-chlorophenyl)-3-oxopropanenitrile |
Specifications
| 4640-66-8 | |
| White-Yellow | |
| MFCD00051625 | |
| 4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag | |
| C1=CC(=CC=C1C(=O)CC#N)Cl | |
| 179.603 | |
| 179.60 | |
| Crystalline Powder |
| 128°C | |
| C9H6ClNO | |
| 5 g | |
| JYOUFPNYTOFCSJ-UHFFFAOYSA-N | |
| 3-(4-chlorophenyl)-3-oxopropanenitrile | |
| 138308 | |
| ≥98.0% (GC) | |
| (4-Chlorobenzoyl)acetonitrile |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator