missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorobenzoyl)acetonitrile 98.0+%, TCI America™

Catalog No. C25825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25825G 5 g
C258225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25825G Supplier TCI America Supplier No. C25825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4640-66-8
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00051625
InChI Key JYOUFPNYTOFCSJ-UHFFFAOYSA-N
Synonym 4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag
PubChem CID 138308
IUPAC Name 3-(4-chlorophenyl)-3-oxopropanenitrile
SMILES C1=CC(=CC=C1C(=O)CC#N)Cl

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 5 g
Formula Weight 179.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (4-Chlorobenzoyl)acetonitrile
TSCA No
Recommended Storage Refrigerator