missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzonitrile, 99%

CAS: 623-03-0 | C7H4ClN | 137.57 g/mol

$263.94 - $263.94

Chemical Identifiers

CAS 623-03-0
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.57
MDL Number MFCD00001813
InChI Key GJNGXPDXRVXSEH-UHFFFAOYSA-N
Synonym p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
PubChem CID 12163
IUPAC Name 4-chlorobenzonitrile
SMILES ClC1=CC=C(C=C1)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151871000
SDS View Documents
Thermo Scientific Chemicals
151871000
100 g Plastic bottle
Each for $263.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

623-03-0
137.57
GJNGXPDXRVXSEH-UHFFFAOYSA-N
12163
ClC1=CC=C(C=C1)C#N
C7H4ClN
MFCD00001813
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
4-chlorobenzonitrile

Specifications

91°C to 94°C
White to Yellow
100.0
100 g
Plastic bottle
98.5% min. (GC)
ClC6H4CN
09, 341
Solubility in water: insoluble
ClC1=CC=C(C=C1)C#N
137.57
137.57
4-Chlorobenzonitrile
623-03-0
98.5
223°C
Authentic
Flakes or Chunks
C7H4ClN
MFCD00001813
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
GJNGXPDXRVXSEH-UHFFFAOYSA-N
4-chlorobenzonitrile
12163
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-765-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only