missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. C012825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C012825G 25 g
C0128500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C012825G Supplier TCI America Supplier No. C012825G

Chemical Identifiers

CAS 98-60-2
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00007439
InChI Key ZLYBFBAHAQEEQQ-UHFFFAOYSA-N
Synonym 4-chlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-chloro, p-chlorophenylsulfonyl chloride, p-chlorobenzenesulfonyl chloride, 4-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, p-chloro, p-chlorbenzensulfochlorid, 4-chlorophenylsulfonyl chloride, 4-chlorobenzenesulphonyl chloride, 4-chloro-benzenesulfonyl chloride
PubChem CID 7398
IUPAC Name 4-chlorobenzene-1-sulfonyl chloride
SMILES ClC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 141°C
Quantity 25 g
UN Number 3261
Formula Weight 211.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzenesulfonyl Chloride
EINECSNumber (3)-1955
RTECSNumber DB8925000
TSCA Yes