missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 158560250

 View more versions of this product

Catalog No. AC158560250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Chlorobenzenesulfonyl chloride
98-60-2
100.0
Authentic
Glass bottle
ClC6H4SO2Cl
25g
4-chlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-chloro, p-chlorophenylsulfonyl chloride, p-chlorobenzenesulfonyl chloride, 4-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, p-chloro, p-chlorbenzensulfochlorid, 4-chlorophenylsulfonyl chloride, 4-chlorobenzenesulphonyl chloride, 4-chloro-benzenesulfonyl chloride
ZLYBFBAHAQEEQQ-UHFFFAOYSA-N
4-chlorobenzenesulfonyl chloride
7398
97%
97%
96.0
107°C
97%
C6H4Cl2O2S
MFCD00007439
11, 55
Solubility in water: insoluble in water
ClC1=CC=C(C=C1)S(Cl)(=O)=O
211.06
211.07

Chemical Identifiers

98-60-2
211.06
ZLYBFBAHAQEEQQ-UHFFFAOYSA-N
7398
ClC1=CC=C(C=C1)S(Cl)(=O)=O
C6H4Cl2O2S
MFCD00007439
4-chlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-chloro, p-chlorophenylsulfonyl chloride, p-chlorobenzenesulfonyl chloride, 4-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, p-chloro, p-chlorbenzensulfochlorid, 4-chlorophenylsulfonyl chloride, 4-chlorobenzenesulphonyl chloride, 4-chloro-benzenesulfonyl chloride
4-chlorobenzenesulfonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-685-3

RTECSNumber : DB8925000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only