missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzamidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America C309025G
Specifications
| 4-Chlorobenzamidine Hydrochloride | |
| 14401-51-5 | |
| 25 g | |
| C7H8Cl2N2 | |
| 4-chlorobenzamidine hydrochloride, 4-chlorobenzene-1-carboximidamide hydrochloride, 4-chlorobenzamidine hcl, p-chlorobenzamidine hydrochloride, 4-chloro-benzamidine hydrochloride, 4-chlorobenzenecarboximidamide monohydrochloride, benzenecarboximidamide, 4-chloro-, monohydrochloride, 4-chlorobenzimidamide hydrochloride, benzamide, p-chloro-, hydrochloride 7ci, benzamidine, p-chloro-, monohydrochloride | |
| C1=CC(=CC=C1C(=N)N)Cl.Cl | |
| 191.055 | |
| 191.06 |
| 246°C | |
| White | |
| Crystal-Powder at 20°C | |
| MFCD00126401 | |
| RXAOGVQDNBYURA-UHFFFAOYSA-N | |
| 4-chlorobenzenecarboximidamide;hydrochloride | |
| 203436 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 14401-51-5 | |
| 191.055 | |
| RXAOGVQDNBYURA-UHFFFAOYSA-N | |
| 203436 | |
| C1=CC(=CC=C1C(=N)N)Cl.Cl |
| C7H8Cl2N2 | |
| MFCD00126401 | |
| 4-chlorobenzamidine hydrochloride, 4-chlorobenzene-1-carboximidamide hydrochloride, 4-chlorobenzamidine hcl, p-chlorobenzamidine hydrochloride, 4-chloro-benzamidine hydrochloride, 4-chlorobenzenecarboximidamide monohydrochloride, benzenecarboximidamide, 4-chloro-, monohydrochloride, 4-chlorobenzimidamide hydrochloride, benzamide, p-chloro-, hydrochloride 7ci, benzamidine, p-chloro-, monohydrochloride | |
| 4-chlorobenzenecarboximidamide;hydrochloride |
Safety and Handling
RTECSNumber : CV6136200
TSCA : No