missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzamidine Hydrochloride 98.0+%, TCI America™

Catalog No. C30905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30905G 5 g
C309025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30905G Supplier TCI America Supplier No. C30905G

Chemical Identifiers

CAS 14401-51-5
Molecular Formula C7H8Cl2N2
Molecular Weight (g/mol) 191.055
MDL Number MFCD00126401
InChI Key RXAOGVQDNBYURA-UHFFFAOYSA-N
Synonym 4-chlorobenzamidine hydrochloride, 4-chlorobenzene-1-carboximidamide hydrochloride, 4-chlorobenzamidine hcl, p-chlorobenzamidine hydrochloride, 4-chloro-benzamidine hydrochloride, 4-chlorobenzenecarboximidamide monohydrochloride, benzenecarboximidamide, 4-chloro-, monohydrochloride, 4-chlorobenzimidamide hydrochloride, benzamide, p-chloro-, hydrochloride 7ci, benzamidine, p-chloro-, monohydrochloride
PubChem CID 203436
IUPAC Name 4-chlorobenzenecarboximidamide;hydrochloride
SMILES C1=CC(=CC=C1C(=N)N)Cl.Cl

Specifications

Melting Point 246°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 191.06
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 4-Chlorobenzamidine Hydrochloride
RTECSNumber CV6136200
TSCA No