missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline, 98%

CAS: 106-47-8 | C6H6ClN | 127.571 g/mol

Supplier:  Thermo Scientific Chemicals A1287930

 View more versions of this product

Catalog No. AAA1287930


Only null left
Explore available promotions

Description

Description

4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.

Solubility
Soluble in water (2.6 g/Liter at 20°C ).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, chloroformates and nitrous acid.
Specifications

Specifications

4-Chloroaniline
106-47-8
232°C
C6H6ClN
UN2018
14,2118
Soluble in water (2.6g/Liter at 20°C ).
C1=CC(=CC=C1N)Cl
127.571
CHEBI:20331
98%
67°C to 72°C
1.43
250 g
MFCD00007835
471359
p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene
QSNSCYSYFYORTR-UHFFFAOYSA-N
4-chloroaniline
7812
127.57

Chemical Identifiers

106-47-8
127.571
QSNSCYSYFYORTR-UHFFFAOYSA-N
7812
4-chloroaniline
C6H6ClN
MFCD00007835
p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene
CHEBI:20331
C1=CC(=CC=C1N)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H350-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c

H301+H311+H331-H317-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID

EINECSNumber : 203-401-0

RTECSNumber : BX0700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only