Learn More
4-Chloroaniline, 98%
CAS: 106-47-8 | C6H6ClN | 127.57 g/mol
Supplier: Thermo Scientific Chemicals 108590010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Chloroaniline | |
| 98% | |
| Beige to Brown-Purple | |
| 100.0 | |
| 1 kg | |
| Glass bottle | |
| 97.5% min. (GC) | |
| ClC6H4NH2 | |
| 1.2 mPa.s (90°C) | |
| 15,2118 | |
| Solubility in water: 3g/L in water (20°C). Other solubilities: freely soluble in alcohol, ether, acetone and, carbon disulfide | |
| C1=CC(=CC=C1N)Cl | |
| 127.57 | |
| CHEBI:20331 | |
| 98% |
| 68.0°C to 72.0°C | |
| 106-47-8 | |
| 97.5 | |
| 232.0°C | |
| Authentic | |
| Crystalline Solid | |
| C6H6ClN | |
| MFCD00007835 | |
| 12,607 | |
| p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene | |
| QSNSCYSYFYORTR-UHFFFAOYSA-N | |
| 4-chloroaniline | |
| 7812 | |
| 127.57 |
Chemical Identifiers
| 106-47-8 | |
| 127.57 | |
| QSNSCYSYFYORTR-UHFFFAOYSA-N | |
| 7812 | |
| 4-chloroaniline |
| C6H6ClN | |
| MFCD00007835 | |
| p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene | |
| CHEBI:20331 | |
| C1=CC(=CC=C1N)Cl |
Safety and Handling
GHS H Statement:
May cause cancer.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-401-0
RTECSNumber : BX0700000
TSCA : TSCA
Recommended Storage : The product turns dark during prolonged storage
RUO – Research Use Only