missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetophenone 97.0+%, TCI America™

Supplier:  TCI America C0033500G

 View more versions of this product

Catalog No. C0033500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4′-Chloroacetophenone
Yellow
C8H7ClO
500 g
4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene
CC(=O)C1=CC=C(Cl)C=C1
154.59
CHEBI:27538
≥97.0% (GC)
99-91-2
237°C
MFCD00000624
3416
BUZYGTVTZYSBCU-UHFFFAOYSA-N
1-(4-chlorophenyl)ethan-1-one
7467
154.59
Liquid

Chemical Identifiers

99-91-2
154.59
BUZYGTVTZYSBCU-UHFFFAOYSA-N
7467
1-(4-chlorophenyl)ethan-1-one
C8H7ClO
MFCD00000624
4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene
CHEBI:27538
CC(=O)C1=CC=C(Cl)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1241

RTECSNumber : KM5600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions