missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetophenone 97.0+%, TCI America™

Catalog No. C003325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C003325G 25 g
C0033100G 100 g
C0033500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C003325G Supplier TCI America Supplier No. C003325G

Chemical Identifiers

CAS 99-91-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000624
InChI Key BUZYGTVTZYSBCU-UHFFFAOYSA-N
Synonym 4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene
PubChem CID 7467
ChEBI CHEBI:27538
IUPAC Name 1-(4-chlorophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 237°C
Quantity 25 g
UN Number 3416
Formula Weight 154.59
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Chloroacetophenone
EINECSNumber (3)-1241
RTECSNumber KM5600000
TSCA Yes