missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetophenone, 98+%

Catalog No. p-7081920
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1211530 250 g
AAA121150B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1211530 Supplier Thermo Scientific Chemicals Supplier No. A1211530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-91-2 | C8H7ClO | 154.59 g/mol

Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals

Solubility
Soluble in water (0.11 mg/ml at 25°C), alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 99-91-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000624
InChI Key BUZYGTVTZYSBCU-UHFFFAOYSA-N
Synonym 4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene
PubChem CID 7467
ChEBI CHEBI:27538
IUPAC Name 1-(4-chlorophenyl)ethanone
SMILES CC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 17°C to 20°C
Density 1.19
Boiling Point 230°C to 232°C
Flash Point 90°C (194°F)
Refractive Index 1.555
Quantity 250 g
UN Number UN3416
Beilstein 386014
Merck Index 14,2116
Solubility Information Soluble in water (0.11mg/ml at 25°C),alcohol,and ether.
Formula Weight 154.6
Percent Purity ≥98%
Chemical Name or Material 4'-Chloroacetophenone
Show More Show Less
Hazard Category H302-H315-H318-H330-H335
Hazard Statement GHS H Statement
H330-H227-H302-H315-H319-H335
Fatal if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, LIQUID
EINECSNumber 202-800-7
RTECSNumber KM5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only