missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetoacetanilide 98.0+%, TCI America™

Catalog No. C062525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C062525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C062525G Supplier TCI America Supplier No. C062525G

Chemical Identifiers

CAS 101-92-8
Molecular Formula C10H10ClNO2
Molecular Weight (g/mol) 211.645
MDL Number MFCD00000613
InChI Key JMRJWEJJUKUBEA-UHFFFAOYSA-N
Synonym 4'-chloroacetoacetanilide, n-4-chlorophenyl-3-oxobutanamide, p-chloroacetoacetanilide, acetoacet-p-chloroanilide, butanamide, n-4-chlorophenyl-3-oxo, acetoacetanilide, 4'-chloro, acetoacetyl-4-chloroanilide, acetoacetanilide, p-chloro, n-4-chlorophenyl acetoacetamide, p-chlorodiacetanilide
PubChem CID 7587
ChEBI CHEBI:35090
IUPAC Name N-(4-chlorophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=C(C=C1)Cl

Specifications

Melting Point 132°C
Color White
Quantity 25 g
Formula Weight 211.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloroacetoacetanilide
EINECSNumber (3)-0220
RTECSNumber AK4375000
TSCA Yes