missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-o-phenylenediamine, 97%

CAS: 95-83-0 | C6H7ClN2 | 142.59 g/mol

$108.21 - $1657.62

Chemical Identifiers

CAS 95-83-0
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00011691
InChI Key BXIXXXYDDJVHDL-UHFFFAOYSA-N
Synonym 4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
PubChem CID 7263
ChEBI CHEBI:82301
IUPAC Name 4-chlorobenzene-1,2-diamine
SMILES NC1=CC=C(Cl)C=C1N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1082314
View Documents
Thermo Scientific Chemicals
A1082314
25 g
Each for $108.21
Only null left
 
AAA1082322
View Documents
Thermo Scientific Chemicals
A1082322
100 g
Each for $353.05
Only null left
 
AAA1082336
View Documents
Thermo Scientific Chemicals
A1082336
500 g
Each for $1,657.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

95-83-0
142.59
BXIXXXYDDJVHDL-UHFFFAOYSA-N
7263
4-chlorobenzene-1,2-diamine
C6H7ClN2
MFCD00011691
4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
CHEBI:82301
NC1=CC=C(Cl)C=C1N

Specifications

70°C to 74°C
25 g
MFCD00011691
BXIXXXYDDJVHDL-UHFFFAOYSA-N
4-chlorobenzene-1,2-diamine
7263
142.59
4-Chloro-o-phenylenediamine
95-83-0
C6H7ClN2
4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
NC1=CC=C(Cl)C=C1N
142.59
CHEBI:82301
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H340-H350
May cause genetic defects.
May cause cancer.

P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335-H350

EINECSNumber : 202-456-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only