missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-o-phenylenediamine, 97%

Catalog No. AAA1082314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1082314 25 g
AAA1082322 100 g
AAA1082336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1082314 Supplier Thermo Scientific Chemicals Supplier No. A1082314

CAS: 95-83-0 | C6H7ClN2 | 142.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-83-0
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00011691
InChI Key BXIXXXYDDJVHDL-UHFFFAOYSA-N
Synonym 4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
PubChem CID 7263
ChEBI CHEBI:82301
IUPAC Name 4-chlorobenzene-1,2-diamine
SMILES NC1=CC=C(Cl)C=C1N

Specifications

Melting Point 70°C to 74°C
Quantity 25 g
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-o-phenylenediamine
Hazard Category H302+H312+H332-H314-H335-H350
Hazard Statement GHS H Statement
H340-H350
May cause genetic defects.
May cause cancer.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
EINECSNumber 202-456-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only