missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-N-methylformanilide 98.0+%, TCI America™

Catalog No. C161925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C161925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C161925G Supplier TCI America Supplier No. C161925G

Chemical Identifiers

CAS 26772-93-0
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.61
MDL Number MFCD02093455
InChI Key XWQPXHSGRKRZPV-UHFFFAOYSA-N
Synonym 4-Chloro-N-formyl-N-methylaniline
PubChem CID 319488
IUPAC Name N-(4-chlorophenyl)-N-methylformamide
SMILES CN(C=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 52°C
Color White
Boiling Point 166°C
Quantity 25 g
Formula Weight 169.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-N-methylformanilide