missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-methylaniline 97.0+%, TCI America™

Catalog No. C16205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16205G 5 g
C162025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16205G Supplier TCI America Supplier No. C16205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 932-96-7
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00000614
InChI Key XCEYKKJMLOFDSS-UHFFFAOYSA-N
Synonym p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine
PubChem CID 70272
IUPAC Name 4-chloro-N-methylaniline
SMILES CNC1=CC=C(C=C1)Cl

Specifications

Boiling Point 142°C
Quantity 5 g
UN Number 1993
Formula Weight 141.60
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Chloro-N-methylaniline
RTECSNumber CX9857900
TSCA No