missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-methylaniline, 95%

CAS: 932-96-7 | C7H8ClN | 141.598 g/mol

$108.92 - $108.92

Chemical Identifiers

CAS 932-96-7
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00000614
InChI Key XCEYKKJMLOFDSS-UHFFFAOYSA-N
Synonym p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine
PubChem CID 70272
IUPAC Name 4-chloro-N-methylaniline
SMILES CNC1=CC=C(C=C1)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2396206
View Documents
Thermo Scientific Chemicals
B2396206
5 g
Each for $108.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

932-96-7
141.598
XCEYKKJMLOFDSS-UHFFFAOYSA-N
70272
CNC1=CC=C(C=C1)Cl
C7H8ClN
MFCD00000614
p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine
4-chloro-N-methylaniline

Specifications

932-96-7
238°C to 240°C
C7H8ClN
MFCD00000614
UN2810
p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine
CNC1=CC=C(C=C1)Cl
141.598
141.6
4-Chloro-N-methylaniline
1.169
>100°C (212°F)
1.584
5 g
2205846
XCEYKKJMLOFDSS-UHFFFAOYSA-N
4-chloro-N-methylaniline
70272
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H318-H335

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 213-262-8

RTECSNumber : CX9857900

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only