missing translation for 'onlineSavingsMsg'
Learn More

Corning™ GL45 Three-Hole Delivery Cap, Red High Temperature (180°C) PBT with PTFE liner

Catalog No. 10462474
Change view
Click to view available options
Thread Size (Metric):
GL 45
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
10462474 GL 45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10462474 Supplier Corning™ Supplier No. 139545DC
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with 1/8 in. O.D. PTFE tubing

These are various sizes and types of screw caps for use with storage bottles.

Order Info

One cap per case

Specifications

Color Red
Material PBT
No. per Case 1/Cs.
No. per Pack 1/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 45
Liner Material PTFE
Neck Finish 39 mm
For Use With (Equipment) Media bottle
Product Type Screw Cap