missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-(4-fluorobenzyl)aniline, 97%, Thermo Scientific™

Catalog No. p-7038088
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56981MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH56981MD Supplier Thermo Scientific Chemicals Supplier No. H56981MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 356531-46-9
Molecular Formula C13H11ClFN
Molecular Weight (g/mol) 235.69
MDL Number MFCD03210761
InChI Key GHZMAHWNLUETDK-UHFFFAOYSA-N
Synonym 4-chloro-n-4-fluorophenyl methyl aniline, 4-chloro-n-4-fluorobenzyl aniline, n-4-fluorobenzyl-4-chloroaniline
PubChem CID 1133333
IUPAC Name 4-chloro-N-[(4-fluorophenyl)methyl]aniline
SMILES FC1=CC=C(CNC2=CC=C(Cl)C=C2)C=C1

Specifications

Quantity 250 mg
Formula Weight 235.69
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(4-fluorobenzyl)aniline
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures