missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-m-phenylenediamine, 97%

Catalog No. AAA1543006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1543006 5 g
AAA1543014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1543006 Supplier Thermo Scientific Chemicals Supplier No. A1543006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5131-60-2 | C6H7ClN2 | 142.586 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5131-60-2
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00025284
InChI Key ZWUBBMDHSZDNTA-UHFFFAOYSA-N
Synonym 4-chloro-m-phenylenediamine, 1,3-benzenediamine, 4-chloro, 4-chloro-1,3-benzenediamine, 4-chloro-1,3-phenylenediamine, 1-chloro-2,4-diaminobenzene, 4-chloro-meta-phenylenediamine, m-phenylenediamine, 4-chloro, 4-chlorophene-1,3-diamine, 4-chloro-1,3-diaminobenzene, 4-chlorophenylene-1,3-diamine
PubChem CID 21209
ChEBI CHEBI:82421
IUPAC Name 4-chlorobenzene-1,3-diamine
SMILES C1=CC(=C(C=C1N)N)Cl

Specifications

Melting Point 87°C to 90°C
Quantity 5 g
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-m-phenylenediamine
Hazard Category H315-H319-H335-H351
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 225-877-9
RTECSNumber SS8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only