missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-D-mandelic Acid 98.0+%, TCI America™
$65.46 - $115.51
Chemical Identifiers
| CAS | 32189-36-9 |
|---|---|
| Molecular Formula | C8H7ClO3 |
| Molecular Weight (g/mol) | 186.591 |
| MDL Number | MFCD00798430 |
| InChI Key | BWSFWXSSALIZAU-SSDOTTSWSA-N |
| Synonym | (R)-2-(4-Chlorophenyl)-2-hydroxyacetic Acid |
| PubChem CID | 785085 |
| IUPAC Name | (2R)-2-(4-chlorophenyl)-2-hydroxyacetic acid |
| SMILES | C1=CC(=CC=C1C(C(=O)O)O)Cl |
Chemical Identifiers
| 32189-36-9 | |
| 186.591 | |
| BWSFWXSSALIZAU-SSDOTTSWSA-N | |
| 785085 | |
| C1=CC(=CC=C1C(C(=O)O)O)Cl |
| C8H7ClO3 | |
| MFCD00798430 | |
| (R)-2-(4-Chlorophenyl)-2-hydroxyacetic Acid | |
| (2R)-2-(4-chlorophenyl)-2-hydroxyacetic acid |
Specifications
| 121°C | |
| White | |
| Crystal-Powder at 20°C | |
| MFCD00798430 | |
| BWSFWXSSALIZAU-SSDOTTSWSA-N | |
| (2R)-2-(4-chlorophenyl)-2-hydroxyacetic acid | |
| 785085 | |
| ≥98.0% (GC,T) |
| 32189-36-9 | |
| 1 g | |
| C8H7ClO3 | |
| (R)-2-(4-Chlorophenyl)-2-hydroxyacetic Acid | |
| C1=CC(=CC=C1C(C(=O)O)O)Cl | |
| 186.591 | |
| 186.59 | |
| 4-Chloro-D-mandelic Acid |
Safety and Handling
TSCA : No