missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-alpha-toluenesulfonyl chloride, 97%

Catalog No. AAH2601706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2601706 5 g
AAH2601703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2601706 Supplier Thermo Scientific Chemicals Supplier No. H2601706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6966-45-6 | C7H6Cl2O2S | 225.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6966-45-6
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
MDL Number MFCD01631927
InChI Key DBJRPJSDYFDWPV-UHFFFAOYSA-N
Synonym 4-chlorophenyl methanesulfonyl chloride, 4-chlorobenzylsulfonyl chloride, 4-chloro-phenyl-methanesulfonyl chloride, 4-chloro-alpha-toluenesulfonyl chloride, chloro 4-chlorophenyl methyl sulfone, 4-chlorobenzenemethanesulfonyl chloride, 4-chlorophenyl methanesulphonyl chloride, 4-chlorophenyl methyl sulfonyl chloride, acmc-20apnh
PubChem CID 227304
IUPAC Name (4-chlorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)Cl

Specifications

Melting Point 93°C to 96°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 225.1
Percent Purity 97%
Chemical Name or Material 4-Chloro-alpha-toluenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only