missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-8-fluoroquinoline 98.0+%, TCI America™

Catalog No. p-7125806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30621G 1 g
C30625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30621G Supplier TCI America Supplier No. C30621G
Only null left

Chemical Identifiers

CAS 63010-72-0
Molecular Formula C9H5ClFN
Molecular Weight (g/mol) 181.59
MDL Number MFCD00278785
InChI Key NUIMKHDPMIPYFI-UHFFFAOYSA-N
PubChem CID 2736587
IUPAC Name 4-chloro-8-fluoroquinoline
SMILES FC1=C2N=CC=C(Cl)C2=CC=C1

Specifications

Melting Point 99°C
Quantity 1 g
Synonym 4-chloro-8-fluoro-quinoline, 8-fluoro-4-chloroquinoline, quinoline, 4-chloro-8-fluoro, pubchem5886, 4-chloro-8fluoroquinoline, 4-chloro-8-fluoroquinolline, quinoline,4-chloro-8-fluoro, 4-chloranyl-8-fluoranyl-quinoline
Formula Weight 181.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-8-fluoroquinoline