missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-(trifluoromethyl)quinoline, 99%

Catalog No. p-7033428
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5053903 1 g
AAH5053906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5053903 Supplier Thermo Scientific Chemicals Supplier No. H5053903

CAS: 49713-56-6 | C10H5ClF3N | 231.602 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49713-56-6
Molecular Formula C10H5ClF3N
Molecular Weight (g/mol) 231.602
MDL Number MFCD00153106
InChI Key FTNQANJWBFKPIP-UHFFFAOYSA-N
Synonym 4-chloro-6-trifluoromethyl quinoline, pubchem5941, 4-chloro-6-trifluoromethyl-quinoline, 4-chloro-6-trifluoromethyl quinolline, quinoline,4-chloro-6-trifluoromethyl
PubChem CID 2736711
IUPAC Name 4-chloro-6-(trifluoromethyl)quinoline
SMILES C1=CC2=NC=CC(=C2C=C1C(F)(F)F)Cl

Specifications

Quantity 1 g
Formula Weight 231.61
Percent Purity 99%
Chemical Name or Material 4-Chloro-6-(trifluoromethyl)quinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only