missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-methylpyrimidine, 97%

Catalog No. p-7055947
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6466703 1 g
AAH6466706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6466703 Supplier Thermo Scientific Chemicals Supplier No. H6466703

CAS: 3435-25-4 | C5H5ClN2 | 128.559 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3435-25-4
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD02322991
InChI Key MVAXKFAQKTWRAH-UHFFFAOYSA-N
Synonym 4-methyl-6-chloro pyrimidine, 4-methyl-6-chloropyrimidine, pyrimidine, 4-chloro-6-methyl, 4-chloro-6-methyl-pyrimidine, 6-chloro-4-methylpyrimidine, 4-chloro-6-methyl-1,3-diazine, pubchem13240, 6-methyl-4-chloropyrimidin, ksc269g3h, pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci
PubChem CID 581796
IUPAC Name 4-chloro-6-methylpyrimidine
SMILES CC1=CC(=NC=N1)Cl

Specifications

Color Yellow
Quantity 1 g
Beilstein 108656
Solubility Information Slightly soluble in water.
Formula Weight 128.56
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-6-methylpyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only