missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-6-methyl-2-methylthiopyrimidine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 17119-73-2 |
|---|---|
| Molecular Formula | C6H7ClN2S |
| Molecular Weight (g/mol) | 174.65 |
| InChI Key | ALMBOXQFPLQVLF-UHFFFAOYSA-N |
| Synonym | 4-chloro-6-methyl-2-methylthio pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-6-methyl-2-methylthio, 4-chloro-6-methyl-2-methylthiopyrimidine, 2-methylmercapto-4-methyl-6-chloropyrimidine, pubchem21461, 6-chloro-4-methyl-2-methylthiopyrimidine, 4-chloro-6-methyl-2-methylthio-pyrimidine, 2-methylthio-4-methyl-6-chloropyrimidine |
| PubChem CID | 241163 |
| IUPAC Name | 4-chloro-6-methyl-2-methylsulfanylpyrimidine |
| SMILES | CC1=CC(=NC(=N1)SC)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC453750010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 17119-73-2 | |
| 174.65 | |
| 4-chloro-6-methyl-2-methylthio pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-6-methyl-2-methylthio, 4-chloro-6-methyl-2-methylthiopyrimidine, 2-methylmercapto-4-methyl-6-chloropyrimidine, pubchem21461, 6-chloro-4-methyl-2-methylthiopyrimidine, 4-chloro-6-methyl-2-methylthio-pyrimidine, 2-methylthio-4-methyl-6-chloropyrimidine | |
| 4-chloro-6-methyl-2-methylsulfanylpyrimidine |
| C6H7ClN2S | |
| ALMBOXQFPLQVLF-UHFFFAOYSA-N | |
| 241163 | |
| CC1=CC(=NC(=N1)SC)Cl |
Specifications
| 17119-73-2 | |
| 96% min. (GC) | |
| C6H7ClN2S | |
| ALMBOXQFPLQVLF-UHFFFAOYSA-N | |
| 4-chloro-6-methyl-2-methylsulfanylpyrimidine | |
| 241163 | |
| 97% |
| Authentic | |
| Glass Bottle | |
| 4-chloro-6-methyl-2-methylthio pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-6-methyl-2-methylthio, 4-chloro-6-methyl-2-methylthiopyrimidine, 2-methylmercapto-4-methyl-6-chloropyrimidine, pubchem21461, 6-chloro-4-methyl-2-methylthiopyrimidine, 4-chloro-6-methyl-2-methylthio-pyrimidine, 2-methylthio-4-methyl-6-chloropyrimidine | |
| CC1=CC(=NC(=N1)SC)Cl | |
| 174.65 | |
| 174.65 | |
| 4-Chloro-6-methyl-2-methylthiopyrimidine |
Safety and Handling
GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only