missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-methoxypyrimidine 98.0+%, TCI America™

$87.60 - $350.44

Chemical Identifiers

CAS 26452-81-3
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.558
MDL Number MFCD06200211
InChI Key KLJGSQVYUGQOAW-UHFFFAOYSA-N
PubChem CID 1051509
IUPAC Name 4-chloro-6-methoxypyrimidine
SMILES COC1=CC(=NC=N1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C23691G
View Documents
TCI America
C23691G
1 g
Each for $87.60
Only null left
 
C23695G
View Documents
TCI America
C23695G
5 g
Each for $350.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

26452-81-3
144.558
KLJGSQVYUGQOAW-UHFFFAOYSA-N
4-chloro-6-methoxypyrimidine
C5H5ClN2O
MFCD06200211
1051509
COC1=CC(=NC=N1)Cl

Specifications

26452-81-3
80°C
MFCD06200211
KLJGSQVYUGQOAW-UHFFFAOYSA-N
4-chloro-6-methoxypyrimidine
1051509
≥98.0% (GC)
4-Chloro-6-methoxypyrimidine
White
C5H5ClN2O
1 g
COC1=CC(=NC=N1)Cl
144.558
144.56
Crystalline Lumps
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'