missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-nitrophthalimide, 95%, Thermo Scientific™

Catalog No. AAL1130803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1130803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1130803 Supplier Thermo Scientific Chemicals Supplier No. L1130803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 6015-57-2
Molecular Formula C8H3ClN2O4
Molecular Weight (g/mol) 226.572
MDL Number MFCD00052331
InChI Key ADLVDYMTBOSDFE-UHFFFAOYSA-N
PubChem CID 2801129
IUPAC Name 5-chloro-6-nitroisoindole-1,3-dione
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])Cl)C(=O)NC2=O

Specifications

Melting Point 198°C to 203°C
Quantity 1 g
Beilstein 1536189
Synonym 4-chloro-5-nitrophthalimide, 5-chloro-6-nitroisoindoline-1,3-dione, 5-chloro-6-nitro-2h-isoindole-1,3-dione, acmc-1b2sd, 5-chloro-6-nitroindoline-1,3-dione, 5-chloro-6-nitro-isoindole-1,3-dione, 6-chloro-5-nitro-2h-benzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,5-chloro-6-nitro, 6-chloro-5-nitro-2h-benzo c azolidine-1,3-dione, 1h-isoindole-1,3 2h-dione, 5-chloro-6-nitro
Formula Weight 226.58
Percent Purity 95%
Chemical Name or Material 4-Chloro-5-nitrophthalimide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures