missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-5-nitro-o-phenylenediamine, 98%

CAS: 67073-39-6 | C6H6ClN3O2 | 187.58 g/mol

$298.08 - $298.08

Chemical Identifiers

CAS 67073-39-6
Molecular Formula C6H6ClN3O2
Molecular Weight (g/mol) 187.58
MDL Number MFCD03427594
InChI Key LOQLMWFVXRZASN-UHFFFAOYSA-N
Synonym 4-chloro-5-nitro-o-phenylenediamine, 4-chloro-5-nitro-1,2-benzenediamine, 5-chloro-4-nitrobenzene-1,2-diamine, 4-chloro-5-nitro-1,2-phenylenediamine, 4-chloro-5-nitro-benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-nitro
PubChem CID 5702618
IUPAC Name 4-chloro-5-nitrobenzene-1,2-diamine
SMILES NC1=CC(Cl)=C(C=C1N)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350660050
SDS View Documents
Thermo Scientific Chemicals
350660050
5 g Glass bottle
Each for $298.08
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67073-39-6
187.58
LOQLMWFVXRZASN-UHFFFAOYSA-N
5702618
NC1=CC(Cl)=C(C=C1N)[N+]([O-])=O
C6H6ClN3O2
MFCD03427594
4-chloro-5-nitro-o-phenylenediamine, 4-chloro-5-nitro-1,2-benzenediamine, 5-chloro-4-nitrobenzene-1,2-diamine, 4-chloro-5-nitro-1,2-phenylenediamine, 4-chloro-5-nitro-benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-nitro
4-chloro-5-nitrobenzene-1,2-diamine

Specifications

67073-39-6
100.0
Red
97.5% min. (HPLC)
C6H6ClN3O2
5 g
LOQLMWFVXRZASN-UHFFFAOYSA-N
4-chloro-5-nitrobenzene-1,2-diamine
5702618
98%
4-chloro-5-nitro-o-phenylenediamine
97.5
189.0°C to 193.0°C
Authentic
Glass bottle
MFCD03427594
4-chloro-5-nitro-o-phenylenediamine, 4-chloro-5-nitro-1,2-benzenediamine, 5-chloro-4-nitrobenzene-1,2-diamine, 4-chloro-5-nitro-1,2-phenylenediamine, 4-chloro-5-nitro-benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-nitro
NC1=CC(Cl)=C(C=C1N)[N+]([O-])=O
187.58
187.59
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only