missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Quantity | 10g |
| Formula Weight | 260.75 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-chloro-5-methyl-6-phenylthieno[2, 3-d]pyrimidine |