missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-methoxy-3(2H)-pyridazinone 98.0+%, TCI America™

Catalog No. C2972200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2972200MG 200 mg
C29721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2972200MG Supplier TCI America Supplier No. C2972200MG

Chemical Identifiers

CAS 63910-43-0
Molecular Formula C5H5ClN2O2
Molecular Weight (g/mol) 160.557
MDL Number MFCD04971392
InChI Key SUZFURZANOKZBL-UHFFFAOYSA-N
PubChem CID 819793
IUPAC Name 5-chloro-4-methoxy-1H-pyridazin-6-one
SMILES COC1=C(C(=O)NN=C1)Cl

Specifications

Melting Point 225°C
Color White-Yellow
Quantity 200 mg
Formula Weight 160.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-5-methoxy-3(2H)-pyridazinone