Learn More
4-chloro-3-nitropyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 429930050
| Quantity | 5g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 13091-23-1 | |
| 158.54 | |
| JOTRPRKONYTVBV-UHFFFAOYSA-N | |
| 2735781 | |
| C1=CN=CC(=C1Cl)[N+](=O)[O-] |
| C5H3ClN2O2 | |
| MFCD02683025 | |
| 4-chloro-3-nitro-pyridine, pyridine, 4-chloro-3-nitro, 3-nitro-4-chloropyridine, 3-nitro-4-chloro-pyridine, zlchem 251, pubchem9253, 4chloro-3-nitropyridine, acmc-1bo4s, 4-chloro-3-nitro pyridine, 3-nitro-4-chloro pyridine | |
| 4-chloro-3-nitropyridine |
Specifications
| 4-Chloro-3-nitropyridine | |
| 96.0 | |
| Authentic | |
| Glass bottle | |
| 5g | |
| 4-chloro-3-nitro-pyridine, pyridine, 4-chloro-3-nitro, 3-nitro-4-chloropyridine, 3-nitro-4-chloro-pyridine, zlchem 251, pubchem9253, 4chloro-3-nitropyridine, acmc-1bo4s, 4-chloro-3-nitro pyridine, 3-nitro-4-chloro pyridine | |
| C1=CN=CC(=C1Cl)[N+](=O)[O-] | |
| 158.54 | |
| 158.54 |
| 13091-23-1 | |
| 100.0 | |
| 97.5% min. (HPLC) | |
| C5H3ClN2O2 | |
| MFCD02683025 | |
| JOTRPRKONYTVBV-UHFFFAOYSA-N | |
| 4-chloro-3-nitropyridine | |
| 2735781 | |
| 97% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear eye protection/face protecti
GHS Signal Word: Danger
Promotions